Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse Thrombopoietin/THPO Animal-Free Protein
SDP

Catalog No. NBP2762611M Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
2 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2762611M 1 mg
NBP2762612U 2 μg
NB27626110U 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP2762611M Supplier Novus Biologicals™ Supplier No. NBP2762611MG

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged bioactive recombinant protein corresponding to Mouse Thrombopoietin/THPO

The Recombinant Mouse Thrombopoietin/THPO Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Gene ID (Entrez) 7066
Molecular Weight (g/mol) 18.8 kDa
Purification Method >95%, by SDS-PAGE
Quantity 1 mg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Gene Symbol THPO
Specific Reactivity Thrombopoietin/THPO is a monomer, 18.8 kDa (175 aa)
Reconstitution Sterile water at 0.1 mg/mL. Upon reconstitution, a small amount of visible precipitate can be expected. A 10% overfill has been added to the total material vialed to compensate for this loss.
Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.