Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse TIM-3 hIgG-His Protein
SDP

Catalog No. NB398091 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398091 500 μg
NB398092 100 μg
NB398090 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB398091 Supplier Novus Biologicals™ Supplier No. NBP307123500UG
Add to Cart
Add to Cart

Recombinant Protein

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4) and 10% Glycerol
Gene ID (Entrez) 84868
Molecular Weight (g/mol) MolecularWeight-theroretical: 46.3 kDa
Quantity 500 μg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Endotoxin Concentration Less Than1 EU / 1 μg of protein (determined by LAL method)
Gene Alias CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3
Gene Symbol HAVCR2
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Form Purified
Research Category Cancer, Immunology, Innate Immunity, Neuroscience
Purity or Quality Grade >95%, by SDS-PAGE
Protein TIM-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.