Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse TRANCE/TNFSF11/RANK L Animal-Free Protein
SDP

Catalog No. NBP2761862U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
2 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2761862U 2 μg
NB27618610U 10 μg
NBP2761861M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP2761862U Supplier Novus Biologicals™ Supplier No. NBP2761862UG

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged bioactive recombinant protein corresponding to Mouse TRANCE/TNFSF11/RANK L

The Recombinant Mouse TRANCE/TNFSF11/RANK L Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Gene ID (Entrez) 8600
Molecular Weight (g/mol) 19.5 kDa
Purification Method >95%, by SDS-PAGE
Quantity 2 μg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Gene Symbol TNFSF11
Specific Reactivity TRANCE/TNFSF11/RANK L is a monomer, 19.5 kDa (175 aa)
Reconstitution Sterile water at 0.1 mg/mL
Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate and 20 mM sodium chloride, pH 7.5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.