Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse VSIG10 Fc Chimera Protein
Click to view available options
Quantity:
50 μg
Description
Measured by its ability to inhibit anti-CD3 antibody induced IL-17 secretion by human peripheral blood mononuclear cells (PBMC). The ED50 for this effect is 0.15-0.9 μg/mL.
Specifications
Specifications
Accession Number | D3YX43 |
Characterization | Bioactivity |
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
Gene ID (Entrez) | 54621 |
Molecular Weight (g/mol) | M.W. (Observed): 97-118 kDa, reducing conditions; M.W. (theoretical): 71 kDa |
Quantity | 50 μg |
Source | Mouse myeloma cell line, NS0-derived mouse VSIG10 protein Mouse VSIG10 (Leu25-Asn426) (N-terminus) Accession # D3YX43 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction