Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Recombinant Plant Amb a 1 Protein

Click to view available options
Quantity:
0.1 mg
Specifications
Specifications
For Use With (Application) | ELISA, SDS-PAGE |
Formulation | Lyophilized from a 0.2 um filtered solution in Storage buffer (100 mM Tris, 150 mM NaCl, 0.01% Tween 20, pH 8.0). |
Quantity | 0.1 mg |
Storage Requirements | Store at -20 to -70C. Avoid freeze-thaw cycles. |
Gene Alias | Ambrosia artemisiifolia 1.0101, Antigen E |
Reconstitution | Reconstitute in sterile deionized water. Use reconstituted product immediately or aliquot for further storage at -20C to -80C. |
Purity or Quality Grade | Multi-step |
Protein | Amb a 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction