Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Recombinant Plant Cup s 1 Protein
SDP

Catalog No. NB158422 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB158422 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB158422 Supplier Novus Biologicals Supplier No. NBP314614
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

For Use With (Application) ELISA, SDS-PAGE
Formulation Lyophilized from a 0.2 um filtered solution in Storage buffer (100 mM Tris, 150 mM NaCl, 0.01% Tween 20, pH 8.0).
Quantity 0.1 mg
Storage Requirements Store at -20 to -70C. Avoid freeze-thaw cycles.
Gene Alias Cupressus sempervirens 1.0101, CUPS1
Reconstitution Reconstitute in sterile deionized water. Use reconstituted product immediately or aliquot for further storage at -20C to -80C.
Purity or Quality Grade Multi-step
Protein Cup s 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.