Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Porcine CXCL8/IL-8 Animal-Free Protein
SDP

Catalog No. NB27618320U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
20 μg
5 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB27618320U 20 μg
NBP2761835U 5 μg
NBP2761831M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB27618320U Supplier Novus Biologicals™ Supplier No. NBP27618320UG

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to Porcine CXCL8/IL-8

The Recombinant Porcine CXCL8/IL-8 Animal-Free Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration Lyoph
For Use With (Application) SDS-PAGE
Gene ID (Entrez) 3576
Molecular Weight (g/mol) 9.1 kDa
Purification Method >95%, by SDS-PAGE
Quantity 20 μg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Gene Symbol CXCL8
Specific Reactivity CXCL8/IL-8 is a monomer, 9.1 kDa (78 aa)
Reconstitution Sterile water at 0.1 mg/mL
Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 5 mM sodium phosphate, 20 mM sodium chloride, pH 7.5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.