Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Protein G Cys (N-Term) Protein
SDP

Catalog No. NB398398 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398398 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398398 Supplier Novus Biologicals™ Supplier No. NBP2349625MG
Add to Cart
Add to Cart

Recombinant Protein

Protein G is a genetically engineered protein containing 3 IgG-binding regions of protein G. Cell wall binding region, cell membrane binding region and albumin binding region have been removed from the recombinant Protein G to ensure the maximum specific IgG binding. The recombinant Protein G is ideal for purification of polyclonal or monoclonal IgG antibodies. Protein G binds to various human, mouse and rat IgG subclasses. It also binds to total IgG from cow, goat, sheep, house and rabbit.

Specifications

For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from additive free solution.
Molecular Weight (g/mol) 21.9 kDa
Quantity 5 mg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 0.1 EU/μg of Protein G as determined by LAL method.
Gene Alias IgG binding protein G Immunoglobulin G binding protein G [Precursor] spg
Reconstitution Dissolve in distilled water or saline.
Research Category Biologically Active Proteins
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein Protein G
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.