Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Recombinant Rat alpha-Synuclein Active, Pre-formed Fibrils Protein
SDP

Catalog No. NB159028 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
2 x 100 μg Vials
5 x 100 μg Vials
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159028 100 μg
NB159029 2 x 100 μg Vials
NB159030 5 x 100 μg Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB159028 Supplier Novus Biologicals Supplier No. NBP314774100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

For Use With (Application) Electron Microscopy, In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot
Formulation PBS pH 7.4
Gene ID (Entrez) 6622
Quantity 100 μg
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein
Gene Symbol SNCA
Research Category Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience
Purity or Quality Grade >95%, by SDS-PAGE
Protein alpha-Synuclein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.