Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Rat CNTFR Protein
SDP

Catalog No. NB253071250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253071250 250 μg
NB25307150 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB253071250 Supplier Novus Biologicals™ Supplier No. NBP253071250UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 23-342 of Rat CNTFR The Recombinant Rat CNTFR Protein is derived from Baculovirus. The Recombinant Rat CNTFR Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 1271
Molecular Weight (g/mol) 36.9kDa
Name Rat CNTFR Protein
Purification Method >90%, by SDS-PAGE
Quantity 250 μg
Source Baculovirus
Immunogen QKHSPQEAPH VQYERLGTDV TLPCGTASWD AAVTWRVNGT DLAPDLLNGS QLILRSLELG HSGLYACFHR DSWHLRHQVL LHVGLPPREP VLSCRSNTYP KGFYCSWHLS APTYIPNTFN VTVLHGSKMM VCEKDPALKN RCHIRYMHLF STIKYKVSIS VSNALGHNTT AITFDEFTIV KPDPPENVVA RPVPSNPRRL EVTWQTPSTW PDPESFPLKF FLRYRPLILD QWQHVELSNG TAHTITDAYA GKEYIIQVAA KDNEIGTWSD WSVAAHATPW TEEPRHLTTE AQAPETTTST TSSLAPPPTT KICDPGELSS LEHHHHHH
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias ciliary neurotrophic factor receptor, ciliary neurotrophic factor receptor alpha, ciliary neurotrophic factor receptor subunit alpha, CNTF receptor subunit alpha, CNTFR alpha, CNTFR-alpha, MGC1774
Gene Symbol CNTFR
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Rat
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.