Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Rat Cyclophilin-F His Protein
SDP

Catalog No. NBP259540X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259540X 100 μg
NBP25954020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP259540X Supplier Novus Biologicals™ Supplier No. NBP259540100UG
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 30-206 of Rat Cyclophilin-F The Recombinant Rat Cyclophilin-F Protein is derived from E. coli. The Recombinant Rat Cyclophilin-F Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 1mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid. PBS (pH7.4) containing 10% glycerol, 1mM DTT
Gene ID (Entrez) 10105
Name Recombinant Rat Cyclophilin-F Protein
Purification Method >90%, by SDS-PAGE
Quantity 100 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias Cyclophilin F, CYP3FLJ90798, Cyp-D, hCyP3, MGC117207, mitochondrial, peptidyl-prolyl cis-trans isomerase, mitochondrial, peptidylprolyl isomerase F, peptidylprolyl isomerase F (cyclophilin F)
Gene Symbol PPIF
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Rat
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.