Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Rat TNF-alpha Protein
SDP

Catalog No. NBP26198620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP26198620 20 μg
NB261986100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP26198620 Supplier Novus Biologicals™ Supplier No. NBP26198620UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity

A Biologically active recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 80-235 of Rat TNF-alpha The Recombinant Rat TNF-alpha Protein is derived from E. coli. The Recombinant Rat TNF-alpha Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration 1 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Phosphate Buffer Saline (pH7.4) containing 10% glycerol, 1mM DTT
Gene ID (Entrez) 7124
Molecular Weight (g/mol) 19.9 kDa
Quantity 20 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1μg of protein (determined by LAL method)
Gene Symbol TNF
Cross Reactivity TNF-alpha
Species Rat
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.