Learn More
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity, SDS-PAGE |
| Formulation | Lyophilized from a 0.2 micron filtered solution, 10 mM sodium phosphate, pH 7.5 |
| Gene ID (Entrez) | 7422 |
| Quantity | 500 μg |
| Storage Requirements | Store at -20 to -70C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 to -70C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Acceptance criteria for endotoxin levels are <1 EUs/μg protein (determined by Kinetic LAL Method). |
| Gene Alias | MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor |
| Gene Symbol | VEGFA |
| Reconstitution | Sterile water at 0.1 mg/mL |
| Research Category | Angiogenesis, Biologically Active Proteins |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
