Learn More
Description
Specifications
Specifications
| Concentration | 0.5mg/mL |
| Formulation | Tris-HCL, 0.2M NaCl, 2mM MgCl2, 0.2mM EDTA, 1M DTT. |
| Gene ID (Entrez) | 5970 |
| Quantity | 5 μg |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO - research use only |
| Recombinant | Recombinant |
| Purity or Quality Grade | Immunogen affinity purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
