Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant SARS-CoV-2 3CL Protease, CF
Click to view available options
Quantity:
50 μg
Description
Recombinant SARS-CoV-2 3CL Protease is a cysteine protease that cleaves proteins with sequences including LQ(S/A/G). The cleavage occurs c-terminal to the glutamine residue. Reaction conditions will need to be optimized for each specific application. We recommend an initial SARS-CoV-2 3CL Protease concentration of 20-40 nM when using substrate S-720.
Specifications
Specifications
Accession Number | YP_009725301.1 |
For Use With (Application) | Bioactivity |
Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl, DTT and EDTA. |
Gene ID (Entrez) | 43740578 |
Molecular Weight (g/mol) | M.W. (theoretical): 34 kDa |
Quantity | 50 μg |
Source | E. coli-derived sars-cov-2 3 CL Protease protein Ser1 - Gln306 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
Conjugate | Unconjugated |
Purity or Quality Grade | >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction