Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant SARS-CoV-2 Envelope N-BBP-His Protein
SDP

Catalog No. NB398143 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398143 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398143 Supplier Novus Biologicals™ Supplier No. NBP307079
Add to Cart
Add to Cart

Recombinant Protein

Specifications

For Use With (Application) Functional Assay
Formulation 0.2 μm filtered solution of 20 mM Tris HCl, 200 mM NaCl, pH 8.0
Gene ID (Entrez) 43740570
Quantity 0.05 mg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias 2019-nCoV Envelope protein, COVID-19 Envelope protein, E Protein, Envelope, Human Coronavirus Envelope protein, SARSCoV2, SARS-CoV-2 E protein, SARSCoV2 Envelope protein, SARS-CoV-2 Envelope protein, Severe acute respiratory syndrome 2 Envelope Protein
Conjugate Unconjugated
Species SARS-CoV-2
Recombinant Recombinant
Form Purified
Purity or Quality Grade >95%, by reducing SDS-PAGE
Protein SARS-CoV-2 Envelope
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.