Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant SARS-CoV-2 nsp13 His (N-Term) Protein
SDP

Catalog No. NB398141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398141 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398141 Supplier Novus Biologicals™ Supplier No. NBP307081
Add to Cart
Add to Cart

Recombinant Protein

Specifications

For Use With (Application) SDS-PAGE
Formulation PBS, pH 7.5, 0.02% NLS
Gene ID (Entrez) 43740578
Molecular Weight (g/mol) MolecularWeight-theroretical: 69.15 kDa
Quantity 0.05 mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Gene Alias helicase, Non-structural protein 13
Conjugate Unconjugated
Species SARS-CoV-2
Recombinant Recombinant
Form Purified
Research Category Infections (Virus Bacteria and Parasites)
Purity or Quality Grade >90%, by SDS-PAGE quantitative densitometry by Coomassie Blue Staining
Protein SARS-CoV-2 nsp13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.