Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant SARS-CoV-2 nsp9 His (N-Term) Protein
SDP

Catalog No. NB151318 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151318 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151318 Supplier Novus Biologicals™ Supplier No. NBP307953
Add to Cart
Add to Cart

Recombinant Protein

Specifications

Formulation ddH2O
Gene ID (Entrez) 1489749
Molecular Weight (g/mol) M.W. Theoretical: 14.67 kDa
Quantity 0.05 mg
Storage Requirements Store at 4 C short term. Store (at -20 C long term. Avoid freeze-thaw cycles.)
Gene Alias Non-structural protein 9, ORF1a polyprotein, ORF1ab polyprotein, p12
Gene Symbol ORF1ab
Purity or Quality Grade >85%, by SDS-PAGE
Protein nsp9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.