Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant SARS-CoV-2 Nucleocapsid His (C-Term) Protein
SDP

Catalog No. NB398138 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398138 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398138 Supplier Novus Biologicals™ Supplier No. NBP307087
Add to Cart
Add to Cart

Recombinant Protein

Specifications

Formulation PBS and 10 mM K2CO3.
Gene ID (Entrez) 43740575
Quantity 0.05 mg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias 2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein
Conjugate Unconjugated
Species SARS-CoV-2
Recombinant Recombinant
Form Purified
Research Category Virology Bacteria and Parasites
Purity or Quality Grade >95% pure by SDS-PAGE
Protein SARS-CoV-2 Nucleocapsid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.