Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant SARS-CoV-2 Nucleocapsid His (C-Term) Protein
SDP

Catalog No. NB025742 Shop All R&D Systems Products
Change view
Click to view available options
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NB025742 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB025742 Supplier Novus Biologicals™ Supplier No. NBP290975
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results

Specifications

Concentration LYOPH
Formulation Lyophilized from PBS and 10 mM K2CO3.
Molecular Weight (g/mol) M.W. (theoretical): 48 kDa
Quantity 50 μg
Storage Requirements Store at -20 C. Avoid freeze-thaw cycles.
Gene Symbol N
Reconstitution Reconstitute in sterile H2O. Recommended concentration is 1.5 mg/mL, but each researcher should determine optimum working concentration for their use.
Purity or Quality Grade >90% SDS-PAGE
Protein SARS-CoV-2 Nucleocapsid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.