Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant SARS-CoV-2 ORF8 His Protein
SDP

Catalog No. NB398136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398136 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398136 Supplier Novus Biologicals™ Supplier No. NBP307089
Add to Cart
Add to Cart

Recombinant Protein

Specifications

For Use With (Application) Western Blot, ELISA, SDS-PAGE
Formulation PBS, pH 7.4
Gene ID (Entrez) 43740577
Molecular Weight (g/mol) MolecularWeight-theroretical: 13.4 kDa
Quantity 0.05 mg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias 2019-nCoV ORF8, 2019-nCoV ORF8 Protein, COVID-19 Non-structural protein 8, COVID-19 ns8, COVID-19 ORF8, Human coronavirus ORF8 Protein, Non-structural protein 8, ns8, ORF8 protein, SARS-CoV-2, SARS-CoV-2 Non-structural protein 8, SARS-CoV-2 ns8, SARSCoV2 ORF8 Protein, SARS-CoV-2 ORF8 Protein, Severe Acute Respiratory Syndrome Coronavirus 2 ORF8 Protein
Conjugate Unconjugated
Species SARS-CoV-2
Recombinant Recombinant
Form Purified
Research Category Infections (Virus Bacteria and Parasites)
Purity or Quality Grade >80% SDS-PAGE
Protein SARS-CoV-2 ORF8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.