Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant SARS-CoV-2 Spike RBD His-tag Avi-tag Protein
Click to view available options
:
50 μg
Description
Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 Fc Chimera (Catalog # 10544-ZN).
Specifications
Specifications
Accession Number | YP_009724390.1 |
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
Molecular Weight (g/mol) | M.W.-observed: 34-41 kDa, under reducing conditions, M.W.-theroretical: 29 kDa |
Quantity | 50 μg |
Source | Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike RBD protein SARS-CoV-2 Spike RBD (Arg319 - Phe541) (N-terminus) Accession # YP_009724390.1 6-His tag Avi-tag (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
Conjugate | Biotin |
Reconstitution | Reconstitute at 500 μg/mL in PBS. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction