Learn More
Novus Biologicals™ Recombinant SARS-CoV-2 Spike S1 (Fc Chimera) His (C-Term) Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Concentration | LYOPH |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | Lyophilized from a 0.22 um filtered solution of PBS, pH 7.4. |
| Gene ID (Entrez) | 43740568 |
| Quantity | 100 μg |
| Storage Requirements | Store at -20 to -70 C. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <0.1 EU/ug of the protein by LAL method. |
| Reconstitution | Reconstitute to a concentration of 0.1-0.5 mg/mL in sterile distilled water. |
| Research Category | Virology Bacteria and Parasites |
| Purity or Quality Grade | >92%, by SDS-PAGE |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.