Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant SARS-CoV 3CL Protease

Catalog No. E718050
missing translation for 'orderingAttributeHoverText'
Quantity:
50 μg
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Carrier Free Recombinant Protein

Recombinant SARS-CoV virus 3CL Protease is a cystein protease that cleaves proteins with sequences including a LQ[S/A/G]. The cleavage occurs c-terminal to the glutamine residue. Reaction conditions will need to be optimized for each specific application.

Specifications

Accession Number P0C6U8
For Use With (Application) Bioactivity
Formulation 1.7 mg/ml (50 μM) in 50 mM HEPES pH 7.5, 150 mM NaCl, 1 mM DTT, 1 mM EDTA
Gene ID (Entrez) 43740578
Molecular Weight (g/mol) M.W.-theoretical: 34 kDa
Quantity 50 μg
Source E. coli-derived sars-cov 3 CL Protease protein Ser3241 - Gln3546
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 60 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Alias 3 CL Proteinase, 3 C-like main protease, 3 CL-Mpro, COVID-19, M Proteinase, Main Protease, Mpro
Conjugate Unconjugated
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein SARS-CoV-2 3 CL Protease
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.