Learn More
Novus Biologicals™ Recombinant SARS-CoV Nucleocapsid His (N-Term) Protein
Shop All R&D Systems Products

Specifications
Specifications
| Formulation | 0.1 M Acetate buffer, pH 4 |
| Quantity | 0.0025 mg |
| Storage Requirements | Store at -20°C to -70°C. Avoid freeze-thaw cycles. |
| Gene Symbol | N |
| Conjugate | Unconjugated |
| Species | SARS-CoV |
| Recombinant | Recombinant |
| Form | Purified |
| Purity or Quality Grade | >95%, by SDS-PAGE |
| Protein | Nucleocapsid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.