Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Sheep IL-13 Protein
SDP

Catalog No. NB397981 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397981 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397981 Supplier Novus Biologicals™ Supplier No. NBP311058
Add to Cart
Add to Cart

Recombinant Protein

Specifications

For Use With (Application) Western Blot, ELISA
Formulation PBS
Gene ID (Entrez) 3596
Molecular Weight (g/mol) MolecularWeight-theroretical: 12.4 kDa
Quantity 5 μg
Storage Requirements Store at -20°C.
Gene Alias ALRHMGC116789, BHR1interleukin-13, IL-13 mgC116788, interleukin 13, MGC116786, NC30, P600
Gene Symbol IL13
Conjugate Unconjugated
Species Sheep
Recombinant Recombinant
Reconstitution Reconstitute with 20 uL sterile PBS containing 0.1% carrier protein. Care should be taken during reconstitution as the protein may appear as a film (at the bottom of the vial. We recommend that the vial is gently mixed after reconstitution.)
Form Purified
Research Category Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Purity or Quality Grade >95%, by SDS-PAGE
Protein IL-13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.