Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Streptolysin Protein
SDP

Catalog No. NB397969 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397969 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397969 Supplier Novus Biologicals™ Supplier No. NBP311627
Add to Cart
Add to Cart

Recombinant Protein

Specifications

For Use With (Application) ELISA
Formulation Tris buffered saline
Quantity 1 mg
Storage Requirements Store at -70°C. Avoid freeze/thaw cycles.
Gene Alias Slo, Thiol activated cytolysin
Conjugate Unconjugated
Species Bacteria
Recombinant Recombinant
Form Purified
Protein Streptolysin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.