Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Viral Monkeypox Virus I1L His (N-Term) Protein
SDP

Catalog No. NB171851
Click to view available options
Quantity:
100 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

Specifications

Concentration LYOPH
For Use With (Application) SDS-PAGE
Formulation Lyophilized from sterile 20mM Tris, 300mM NaCl, 1mM DTT, 50% Glycerol, pH 7.4.
Molecular Weight (g/mol) M.W. Theoretical: 37.71 kDa
Quantity 100 μg
Storage Requirements Store at -20°C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <1.0 EU per ug protein as determined by the LAL method.
Reconstitution Reconstitution instructions are lot-specific, please contact technical support
Purity or Quality Grade >90% SDS-PAGE
Protein Monkeypox Virus I1L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.