Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Viral Yellow Fever Virus NS1 Protein
Click to view available options
Quantity:
100 μg
Description
The Recombinant Viral Yellow Fever Virus NS1 Protein is derived from HEK293.
Specifications
Specifications
Accession Number | P03314 |
For Use With (Application) | Flow Cytometry |
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Molecular Weight (g/mol) | 41 kDa |
Quantity | 100 μg |
Source | Human embryonic kidney cell,HEK293-derived viral Yellow Fever Virus NS1 protein Gln780-His1134,with C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
Conjugate | Unconjugated |
Species | Virus |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction