Learn More
Novus Biologicals Recombinant Virus Chikungunya Virus E1 His (C-Term) Protein
Shop All R&D Systems Products

Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl pH 7.8, 210mM NaCl |
| Quantity | 0.5 mg |
| Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
| Gene Alias | Chikungunya Virus |
| Purity or Quality Grade | >95% pure by SDS-PAGE |
| Protein | Chikungunya Virus E1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.