Learn More
Specifications
Specifications
| For Use With (Application) | Electron Microscopy, Western Blot |
| Formulation | 10mM sodium phosphate, 20mM sodium citrate, 154mM sodium chloride, pH7.4 |
| Quantity | 0.25 mg |
| Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
| Gene Alias | polyprotein precursor |
| Purity or Quality Grade | >95% pure by SDS-PAGE |
| Protein | Dengue Virus 1 VLP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
