Learn More
Novus Biologicals Recombinant Virus Hepatitis E Virus Capsid His (C-Term) Protein
Shop All R&D Systems Products

Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | DPBS pH7.4 |
| Molecular Weight (g/mol) | theroretical M.W.: 70 kDa |
| Quantity | 0.5 mg |
| Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
| Gene Alias | ORF2, pORF2, Secreted protein ORF2 |
| Purity or Quality Grade | >95% |
| Protein | Hepatitis E Virus Capsid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.