Learn More
Novus Biologicals Recombinant Virus Yellow Fever Virus Envelope Fc Protein
Shop All R&D Systems Products

Specifications
Specifications
| For Use With (Application) | ELISA |
| Formulation | DPBS pH 7.4 |
| Quantity | 0.1 mg |
| Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
| Gene Alias | Envelope protein E, polyprotein, partial, YFV E, YFV Envelope protein |
| Purity or Quality Grade | >90% |
| Protein | Yellow Fever Virus Envelope |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.