Learn More
Novus Biologicals Recombinant Virus Zika Virus Envelope Suriname Z1106033 His (C-Term) Protein
Shop All R&D Systems Products

Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM NaH2PO4 pH 7.8, 210mM NaCl |
| Quantity | 0.1 mg |
| Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
| Gene Alias | polyprotein, partial |
| Purity or Quality Grade | >95% |
| Protein | Zika Virus Envelope |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.