Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Recombinant Virus Zika Virus VLP Suriname Z1106033 Protein
SDP

Catalog No. NB159147 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159147 0.25 mg
NB159146 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB159147 Supplier Novus Biologicals Supplier No. NBP3148350.25MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

For Use With (Application) SDS-PAGE
Formulation DPBS pH 7.4, 20% Sucrose. May contain traces of tartrate
Quantity 0.25 mg
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias polyprotein, partial
Purity or Quality Grade >95% pure by SDS-PAGE
Protein Zika Virus VLP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.