Learn More
Novus Biologicals™ Recombinant Western Blot Protein Standards (12-90 kDa)

Specifications
Specifications
| Buffer | 125 mM Tris-HCl, pH 6.8, 10 mM DTT, 2 mM EDTA, 2 % (W/V) SDS, 10 % (W/V) Glycerol, 0.01% (W/V) Bromophenol blue |
| Content And Storage | Store at -20°C. |
| For Use With (Application) | Western Blot |
| Quantity | 250 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.