Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Yeast AHSA1 His Protein
SDP

Catalog No. NB172001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172001 100 μg
NB172003 50 μg
NB172002 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB172001 Supplier Novus Biologicals™ Supplier No. NBP318327100UG
This item is not returnable. View return policy

Recombinant Protein

Specifications

For Use With (Application) SDS-PAGE, Western Blot
Formulation 12.5mM Hepes buffer pH7.2, 10mM NaCl, 50% glycerol
Gene ID (Entrez) 10598
Molecular Weight (g/mol) M.W. Theoretical: 405 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1
Gene Symbol AHSA1
Research Category Signal Transduction
Purity or Quality Grade >85%
Protein AHSA1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.