Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RecoverAll™ Total Nucleic Acid Isolation Kit for FFPE

Catalog No. am1975
Encompass_Preferred
Click to view available options
Quantity:
40 Preps

Includes

16mL Digestion Buffer; 60mL Wash 1 Concentrate; 60mL Wash 2/3 Concentrate; 80 Collection Tubes; 40 Filter Cartridges; 19.2mL Isolation Additive; 5mL Elution Solution; 160μL Protease; 240μL 10X DNase Buffer; 160μL DNase; 400μL RNase A

The RecoverAll™ Total Nucleic Acid Isolation Kit for FFPE is for the extraction of total nucleic acid from formalin- or paraformalin-fixed, paraffin-embedded (FFPE) tissues.

The RecoverAll™ Total Nucleic Acid Isolation Kit for FFPE is for the extraction of total nucleic acid from formalin- or paraformalin-fixed, paraffin-embedded (FFPE) tissues. Sufficient reagents are included for 40 purifications from up to four 20 μm sections, or up to 35 mg of unsectioned, core samples each. Features of the RecoverAll™ Total Nucleic Acid Isolation Kit for FFPE:

• Optimized for isolation of total nucleic acids, including microRNAs, from FFPE tissue
• No overnight Proteinase K digestion required—deparaffinize in the morning and perform qRT-PCR in the afternoon
• Typical yields are >50% of those of unfixed tissue
• Recovered nucleic acids are suitable for next generation sequencing, real-time RT-PCR, PCR, mutation screening, and microarray analyses

Extraction of nucleic acids from difficult samples
Archived tissue samples contain valuable information of disease states, but it has traditionally been difficult to isolate nucleic acids from them of a quality suitable for molecular analysis. Standard preservation techniques use formalin that maintains tissue structure and prevents putrefaction, but which also traps nucleic acids and modifies them through protein-protein and protein-nucleic acid crosslinks. RNA (and to some extent DNA) is often so fragmented and chemically modified that it is incompatible with many molecular analysis techniques. RNA fragmentation in FFPE tissues cannot be reversed; however, the protease digestion conditions of the RecoverAll™ kit are designed to release a maximal amount (see figure) of trapped RNA fragments of all sizes, including microRNA, in a relatively short amount of time.

The RecoverAll™ Total Nucleic Acid Isolation Kit solution
The RecoverAll™ Total Nucleic Acid Isolation Kit procedure requires about 45 minutes of hands-on time and can be completed in typically less than 1 day for RNA. FFPE samples are deparaffinized using a series of xylene and ethanol washes. Next, they are subjected to an extensive protease digestion with an incubation time tailored for the recovery of either RNA or DNA. Nucleic acids are then purified using a rapid glass-filter method, and are eluted into either water or low-salt buffer.

Nucleic acids for almost any downstream application
As is the case with all FFPE tissue, sample fixation and storage typically cause nucleic acid fragmentation and modification. Therefore, downstream applications, such as microarray analysis, which require more pristine RNA than does qRT-PCR, may require modification for optimal results. Although DNA tends not to fragment as easily as RNA, it appears to be more reactive to the formalin and requires a longer (2-day) protease digestion time to release substantial amounts of DNA.

Not high-throughput compatible (Manual)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

Box 1
• 16 mL Digestion Buffer; room temperature
• 60 mL Wash 1 (concentrated); room temperature
• 60 mL Wash 2/3 (concentrated); room temperature
• 80 Collection Tubes; room temperature
• 40 Filter Cartridges; room temperature
• 19.2 mL Isolation Additive; room temperature
• 5 mL Elution Solution; room temperature

Box 2
• 160 μL Protease; -20°C
• 240 μL 10X DNase Buffer; -20°C
• 160 μL DNase; -20°C
• 400 μL RNase A; -20°C

Isolation Technology Spin Column (Glass Fiber Filter)
Purification Time 45 min. (Hands-on)
Elution Volume 60 μL
Sample Type FFPE Tissue, Paraffin-embedded (FFPE) Tissue
Final Product Type Total RNA and DNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization
Quantity 40 Preps
Shipping Condition Box 1: Room Temperature
Box 2: Dry Ice
Starting Material Amount ≤80 μM tissue slices
35 mg unsectioned core
Yield DNA: ≤3.5 μg per mg FFPE
RNA: varies
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.