Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Recoverin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP33825820UL
Description
Recoverin Polyclonal antibody specifically detects Recoverin in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ImmunofluorescenceSpecifications
Recoverin | |
Polyclonal | |
Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
cancer associated retinopathy antigen, Protein CAR, RCV1Cancer-associated retinopathy protein, recoverin | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human Recoverin (NP_002894.1).,, Sequence:, MGNSKSGALSKEILEELQLNTKFSEEELCSWYQSFLKDCPTGRITQQQFQSIYAKFFPDTDPKAYAQHVFRSFDSNLDGTLDFKEYVIALHMTTAGKTNQKLEWAFSLYDVDGNGTISKNEVLEIVMAIFKMITPEDVKLLPDDENTPEKRAEKIWKYFGKNDDDKLTEKEFIEGTLANKEILRLIQFEPQKVKEKMKNA | |
20 μL | |
Neuroscience, Vision | |
5957 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
Unconjugated | |
PBS (pH 7.3), 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction