Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Recoverin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 3B8E8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | Recoverin |
|---|---|
| Clone | 3B8E8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6942
![]()
|
Proteintech Group Inc
665211IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Seems to be implicated in the pathway from retinal rod guanylate cyclase to rhodopsin. May be involved in the inhibition of the phosphorylation of rhodopsin in a calcium-dependent manner. The calcium-bound recoverin prolongs the photoresponse.Specifications
| Recoverin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Protein CAR, RCV1, RCVRN, recoverin | |
| RCVRN | |
| IgG1 | |
| Protein G | |
| Antibody |
| 3B8E8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Pig, Rat | |
| P34057, P35243 | |
| 100516153, 140936, 19674, 5957 | |
| Recoverin Fusion Protein Ag0119 | |
| Primary | |
| -20°C | |
| RCVRN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title