Learn More
Invitrogen™ Recoverin Recombinant Rabbit Monoclonal Antibody (4G6B4)
Rabbit Recombinant Monoclonal Antibody
$169.00 - $419.00
Specifications
Antigen | Recoverin |
---|---|
Clone | 4G6B4 |
Concentration | 0.21 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Western Blot |
Description
Immunogen sequence: MGNSKSGALS KEILEELQLN TKFSEEELCS WYQSFLKDCP TGRITQQQFQ SIYAKFFPDT DPKAYAQHVF RSFDSNLDGT LDFKEYVIAL HMTTAGKTNQ KLEWAFSLYD VDGNGTISKN EVLEIVMAIF KMITPEDVKL LPDDENTPEK RAEKIWKYFG KNDDDKLTEK EFIEGTLANK EILRLIQFEP QKVKEKMKNA.
Seems to be implicated in the pathway from retinal rod guanylate cyclase to rhodopsin. May be involved in the inhibition of the phosphorylation of rhodopsin in a calcium-dependent manner. The calcium-bound recoverin prolongs the photoresponse.Specifications
Recoverin | |
0.21 mg/mL | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
P35243 | |
5957 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human Recoverin (NP_002894.1). | |
Antibody |
4G6B4 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
RCVRN | |
23 kDa photoreceptor cell-specific protein; cancer associated retinopathy antigen; cancer associated retinopathy protein; cancer-associated retinopathy protein; CAR; guanylate cyclase activator; p26; P26-calcium binding protein; Protein CAR; RCV1; RCVRN; Recoverin; S-modulin | |
RCVRN | |
Primary | |
Protein A |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.