Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RECQ1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299352MFV450
Description
RECQ1 Polyclonal antibody specifically detects RECQ1 in Human samples. It is validated for Flow CytometrySpecifications
RECQ1 | |
Polyclonal | |
Flow Cytometry | |
DNA helicase, RecQ-like type 1, DNA-dependent ATPase Q1, EC 3.6.1, EC 3.6.4.12, RecQ protein-like (DNA helicase Q1-like), RecQ protein-like 1, RECQ1, RecQ1ATP-dependent DNA helicase Q1, RecQL1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human RECQ1. | |
0.1 mL | |
DNA Repair | |
5965 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction