Learn More
Invitrogen™ RECQ1 Monoclonal Antibody (4D5)

Description
Peptide Sequence: FLIQQYLKED YSFTAYATIS YLKIGPKANL LNNEAHAITM QVTKSTQNSF RAESSQTCHS EQGDKKMEEK NSGNFQKKAA NMLQQSGSKN TGAKKRKIDD.
Specifications
Specifications
| Antigen | RECQ1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4D5 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RECQL |
| Gene Accession No. | P46063 |
| Gene Alias | ATP-dependent DNA helicase Q1; DNA helicase, RecQ-like type 1; DNA-dependent ATPase Q1; RecQ helicase-like; RecQ like helicase; RecQ protein-like; RecQ protein-like (DNA helicase Q1-like); recQ protein-like 1; RecQ1; RECQL; RECQL1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.