Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RecQ1 Mouse anti-Human, Unconjugated, Clone: 5A12.1, MilliporeSigma™

Catalog No. MABE1055100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1055-100 100 μL
MABE1055-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE1055-100 Supplier MilliporeSigma Supplier No. MABE1055100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-RecQ1, clone 5A12.1, Cat. No. MABE1055, is a highly specific mouse monoclonal antibody that targets RecQ1 and has been tested for use in Western Blotting and Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RecQ1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5A12.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias ATP-dependent DNA helicase Q1;EC:3.6.4.12;DNA helicase;RecQ-like type 1;RecQ1;DNA-dependent ATPase Q1;RecQ protein-like 1
Gene Symbols RECQL;RECQ1;RECQL1
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 87 amino acids from the internal region of human ATP-dependent DNA helicase Q1.
Molecular Weight of Antigen ∽73 kDa observed; 73.46 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_002898
Test Specificity Clone 5A12.1 specifically detects human ATP-dependent DNA helicase Q1 (RecQ1). It targets an epitope within 87 amino acids from the internal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.