Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RECQ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583169 Supplier Invitrogen™ Supplier No. PA583169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HVLTDAQKIL CIEKCFTFTA SFNRPNLYYE VRQKPSNTED FIEDIVKLIN GRYKGQSGII YCFSQKDSEQ VTVSLQNLGI HAGAYHANLE PEDK Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 83%.

The protein encoded by this gene is a member of the RecQ DNA helicase family. DNA helicases are enzymes involved in various types of DNA repair, including mismatch repair, nucleotide excision repair and direct repair. Some members of this family are associated with genetic disorders with predisposition to malignancy and chromosomal instability. The biological function of this helicase has not yet been determined. Two alternatively spliced transcripts, which encode the same isoform but differ in their 5' and 3' UTRs, have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RECQ1
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RECQL
Gene Accession No. P46063
Gene Alias ATP-dependent DNA helicase Q1; DNA helicase, RecQ-like type 1; DNA-dependent ATPase Q1; RecQ helicase-like; RecQ like helicase; RecQ protein-like; RecQ protein-like (DNA helicase Q1-like); recQ protein-like 1; RecQ1; RECQL; RECQL1
Gene Symbols RECQL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RECQL. Recombinant protein control fragment (Product #RP-95478).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5965
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.