Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RECQ4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555263
Description
Immunogen sequence: WLQRCHSEVP DFLGAPKACR PDLGSEESQL LIPGESAVLG PGAGSQGPEA SAFQEVSIRV GSPQPSSSGG EKRRWNEEPW ESPAQVQQES SQAGPPSEGA GAVAVEEDPP GE Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 45%.
The protein encoded by this gene is a DNA helicase that belongs to the RecQ helicase family. DNA helicases unwind double-stranded DNA into single-stranded DNAs and may modulate chromosome segregation. This gene is predominantly expressed in thymus and testis. Mutations in this gene are associated with Rothmund-Thomson, RAPADILINO and Baller-Gerold syndromes.
Specifications
RECQ4 | |
Polyclonal | |
Unconjugated | |
RECQL4 | |
ATP-dependent DNA helicase Q4; DNA helicase, RecQ-like type 4; DNA helicase, RecQ-like, type 4; RecQ helicase protein-like 4; RecQ helicase-like 4; RecQ like helicase 4; RecQ protein-like 4; Recq4; RECQL4; RTS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9401 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94761 | |
RECQL4 | |
Recombinant protein corresponding to Human RECQ4. Recombinant protein control fragment (Product #RP-89489). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction