Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RECQ4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593245 Supplier Invitrogen™ Supplier No. PA593245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAGSQGPEAS AFQEVSIRVG SPQPSSSGGE KRRWNEEPWE SPAQVQQESS QAGPPSEGAG AVAVEEDPPG EPVQAQPPQP CSSPSNPRYH GLSPSSQARA; Positive Samples: HeLa, Mouse thymus; Cellular Location: Cytoplasm, Nucleus.

The protein encoded by this gene is a DNA helicase that belongs to the RecQ helicase family. DNA helicases unwind double-stranded DNA into single-stranded DNAs and may modulate chromosome segregation. This gene is predominantly expressed in thymus and testis. Mutations in this gene are associated with Rothmund-Thomson, RAPADILINO and Baller-Gerold syndromes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RECQ4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.313 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RECQL4
Gene Accession No. O94761, Q75NR7
Gene Alias ATP-dependent DNA helicase Q4; DNA helicase, RecQ-like type 4; DNA helicase, RecQ-like, type 4; RecQ helicase protein-like 4; RecQ helicase-like 4; RecQ like helicase 4; RecQ protein-like 4; Recq4; RECQL4; RTS
Gene Symbols RECQL4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 231-330 of human RECQL4 (NP_0042513).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79456, 9401
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.