Learn More
Description
Specifications
Specifications
| Antigen | RECQL5 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000-1:10000, Immunoprecipitation 5-15 ug/mg lysate |
| Formulation | TBS and 0.1% BSA with 0.09% Sodium Azide |
| Gene Alias | DNA helicase, RecQ-like type 5, EC 3.6.1, EC 3.6.4.12, FLJ90603, RecQ protein-like 5ATP-dependent DNA helicase Q5, RecQ5, RECQ5RecQ protein 5 |
| Gene Symbols | RECQL5 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
