Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RECQL5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556315
Description
Immunogen sequence: ELEHETFRNA KVANLYKASV LKKVADIHRA SKDGQPYDMG GSAKSCSAQA EPPEPNEYDI PPASHVYSLK PKRVGAGFPK GSCPFQTATE LMETTRIREQ Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.
RECQL5 gene ontology annotations related to this gene include cellular response to camptothecin; chromosome separation; DNA duplex unwinding; DNA recombination; DNA repair; DNA replication; double-strand break repair via homologous recombination; mitotic sister chromatid segregation; negative regulation of double-strand break repair via homologous recombination; negative regulation of mitotic cell cycle; negative regulation of transcription elongation from RNA polymerase II promoter; replication-born double-strand break repair via sister chromatid exchange; response to X-ray.
Specifications
RECQL5 | |
Polyclonal | |
Unconjugated | |
RECQL5 | |
ATP-dependent DNA helicase Q5; DNA helicase, RecQ-like type 5; FLJ90603; LOW QUALITY PROTEIN: ATP-dependent DNA helicase Q5; RecQ helicase-like 5; RecQ like helicase 5; RecQ protein 5; RecQ protein-like 5; RecQ protein-like 5 like; RECQ5; Recq5b; RECQL5; Recql5b; RECQL5beta; recql5l | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9400 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94762 | |
RECQL5 | |
Recombinant protein corresponding to Human RECQL5. Recombinant protein control fragment (Product #RP-95484). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction