Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RECQL5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981190
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1190 Supplier Bioss Supplier No. BS11231R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RECQL5 gene ontology annotations related to this gene include cellular response to camptothecin; chromosome separation; DNA duplex unwinding; DNA recombination; DNA repair; DNA replication; double-strand break repair via homologous recombination; mitotic sister chromatid segregation; negative regulation of double-strand break repair via homologous recombination; negative regulation of mitotic cell cycle; negative regulation of transcription elongation from RNA polymerase II promoter; replication-born double-strand break repair via sister chromatid exchange; response to X-ray.

Specifications

Antigen RECQL5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RECQL5
Gene Accession No. O94762
Gene Alias ATP-dependent DNA helicase Q5; BOS_19480; DNA helicase, RecQ-like type 5; FLJ90603; RecQ helicase-like 5; RecQ like helicase 5; RecQ protein 5; RecQ protein-like 5; RECQ5; Recq5b; Recql5; Recql5b; RECQL5beta; recql5l
Gene Symbols RECQL5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RECQL5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9400
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.